Learn More
1,4,7,14,17,20-Hexaoxacyclohexacosane-8,13,21,26-tetrone, TRC
Product Code. 30406787
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30406787 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30406787 Supplier Toronto Research Chemicals Supplier No. TRCH28493050MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1,4,7,14,17,20-Hexaoxacyclohexacosane-8,13,21,26-tetrone
CAS 314254-68-7
Molecular Formula C20 H32 O10
Quantity 50 mg
Synonym 4-(Dimethylamino)-1-[1-[3-(dimethylamino)propyl]-1-(4-fluorophenyl)-1,3-dihydro-5-isobenzofuranyl]-1-butanone
InChI Formula InChI=1S/C20H32O10/c21-17-5-1-2-6-18(22)28-14-10-26-12-16-30-20(24)8-4-3-7-19(23)29-15-11-25-9-13-27-17/h1-16H2
SMILES O=C1CCCCC(=O)OCCOCCOC(=O)CCCCC(=O)OCCOCCO1
IUPAC Name 1,4,7,14,17,20-hexaoxacyclohexacosane-8,13,21,26-tetrone
Molecular Weight (g/mol) 432.462
Formula Weight 432.2