Learn More
1,4,7,14,17,20-Hexaoxacyclohexacosane-8,13,21,26-tetrone, TRC
Specifications
Specifications
| Chemical Name or Material | 1,4,7,14,17,20-Hexaoxacyclohexacosane-8,13,21,26-tetrone |
| CAS | 314254-68-7 |
| Molecular Formula | C20 H32 O10 |
| Quantity | 5 mg |
| Synonym | 4-(Dimethylamino)-1-[1-[3-(dimethylamino)propyl]-1-(4-fluorophenyl)-1,3-dihydro-5-isobenzofuranyl]-1-butanone |
| InChI Formula | InChI=1S/C20H32O10/c21-17-5-1-2-6-18(22)28-14-10-26-12-16-30-20(24)8-4-3-7-19(23)29-15-11-25-9-13-27-17/h1-16H2 |
| SMILES | O=C1CCCCC(=O)OCCOCCOC(=O)CCCCC(=O)OCCOCCO1 |
| IUPAC Name | 1,4,7,14,17,20-hexaoxacyclohexacosane-8,13,21,26-tetrone |
| Molecular Weight (g/mol) | 432.462 |
| Formula Weight | 432.2 |