Learn More
1,3-Dipalmitoyl-2-oleoyl Glycerol-d5, TRC
Specifications
Specifications
| Chemical Name or Material | 1,3-Dipalmitoyl-2-oleoyl Glycerol-d5 |
| CAS | 2734692-62-5 |
| Molecular Formula | C53 H95 D5 O6 |
| Quantity | 1 mg |
| Synonym | (9Z)-9-Octadecenoic Acid 2-[(1-Oxohexadecyl)oxy]-1-[[(1-oxohexadecyl)oxy]methyl]ethyl Ester-d5, 1,3-Dipalmito-2-olein-d5, 1,3-Dipalmitoyl-2-olein-d5, 1,3-Dipalmitoyl-2-oleoylglycerin-d5, 1,3-Dipalmitoyl-2-oleoylglycerol-d5, 1-Palmito-2-oleo-3-palmitin-d5, 2-Oleo-1,3-dipalmitin-d5, 2-Oleodipalmitin-d5, 2-Oleoyl-1,3-dipalmitin-d5, 2-Oleoyl-1,3-dipalmitoylglycerin-d5, 2-Oleoyl-1,3-dipalmitoylglycerol-d5, Glycerin-1-palmitate-2-oleate-3-palmitate-d5, Glycerol 1,3-Dipalmitate 2-oleate-d5, Glyceryl 2-oleate-1,3-dipalmitate-d5, POP-d5 |
| InChI Formula | InChI=1S/C53H100O6/c1-4-7-10-13-16-19-22-25-26-29-32-35-38-41-44-47-53(56)59-50(48-57-51(54)45-42-39-36-33-30-27-23-20-17-14-11-8-5-2)49-58-52(55)46-43-40-37-34-31-28-24-21-18-15-12-9-6-3/h25-26,50H,4-24,27-49H2,1-3H3/i48D2,49D2,50D |
| SMILES | C(OC(CCCCCCCC=CCCCCCCCC)=O)(C(OC(CCCCCCCCCCCCCCC)=O)([2H])[2H])(C(OC(CCCCCCCCCCCCCCC)=O)([2H])[2H])[2H] |
| IUPAC Name | [1,2,2-trideuterio-1-[dideuterio(hexadecanoyloxy)methyl]-2-hexadecanoyloxy-ethyl] octadec-9-enoate |
| Molecular Weight (g/mol) | 838.38831 |
| Formula Weight | 837.7834 |