missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dichloro-4-fluorobenzene, 99%, ACROS Organics™

Product Code. 10144730 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
Unit Size:
25mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10144730 5 mL 5mL
10561121 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10144730 Supplier Thermo Scientific Acros Supplier No. 270630050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 1435-48-9
Molecular Formula C6H3Cl2F
Molecular Weight (g/mol) 164.988
MDL Number MFCD00000580
InChI Key BDJZCCWUSOZUQG-UHFFFAOYSA-N
Synonym 1,3-dichloro-4-fluorobenzene, 2,4-dichlorofluorobenzene, benzene, 2,4-dichloro-1-fluoro, dcfb, benzene,2,4-dichloro-1-fluoro, 2,4-dichloro-1-fluoro-benzene, pubchem2312, acmc-209cqu, 2,4-dichlorofluobenzene, 1,3-dichlorofluorobenzene
PubChem CID 123112
IUPAC Name 2,4-dichloro-1-fluorobenzene
SMILES C1=CC(=C(C=C1Cl)Cl)F

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.409g/mL
Infrared Spectrum Authentic
Assay Percent Range 99%
Linear Formula Cl2C6H3F
Refractive Index 1.5235 to 1.5255 (20°C, 589nm)
Quantity 5 mL
Beilstein 05, 547
Formula Weight 164.99
Percent Purity ≥98.5%
Physical Form Liquid
Chemical Name or Material 1, 3-Dichloro-4-fluorobenzene, 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.