missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dicaffeoylquinic acid, MedChemExpress

Product Code. p-200101262
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16483916 25 mg 25mg
16459218 10 mg 10mg
16401638 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16483916 Supplier Medchem Express Supplier No. HYN1412
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress 1,3-Dicaffeoylquinic acid is a caffeoylquinic acid derivative that exhibits antioxidant activity and radical scavenging activity.

1,3-Dicaffeoylquinic acid is a caffeoylquinic acid derivative that exhibits antioxidant activity and radical scavenging activity.

Specifications

Chemical Name or Material 1,3-Dicaffeoylquinic acid
CAS 19870-46-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C25H24O12
Quantity 25 mg
Synonym 1,3-O-Dicaffeoylquinic acid 1,5-Dicaffeoylquinic acid
Solubility Information DMSO : 100 mg/mL (193.63 mM; Need ultrasonic)
SMILES O=C([C@@]1(OC(/C=C/C2=CC=C(O)C(O)=C2)=O)C[C@@H](OC(/C=C/C3=CC=C(O)C(O)=C3)=O)[C@H](O)[C@H](O)C1)O
Molecular Weight (g/mol) 516.45
Formula Weight 516.45
Percent Purity 98.68%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.