Learn More
1,3-Dicaffeoylquinic acid, MedChemExpress
Product Code. 16483916
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16483916 25 mg 25mg
16459218 10 mg 10mg
16401638 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16483916 Supplier Medchem Express Supplier No. HYN1412
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
MedChemExpress 1,3-Dicaffeoylquinic acid is a caffeoylquinic acid derivative that exhibits antioxidant activity and radical scavenging activity.
1,3-Dicaffeoylquinic acid is a caffeoylquinic acid derivative that exhibits antioxidant activity and radical scavenging activity.
Specifications
Chemical Name or Material 1,3-Dicaffeoylquinic acid
CAS 19870-46-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C25H24O12
Quantity 25 mg
Synonym 1,3-O-Dicaffeoylquinic acid 1,5-Dicaffeoylquinic acid
Solubility Information DMSO : 100 mg/mL (193.63 mM; Need ultrasonic)
SMILES O=C([C@@]1(OC(/C=C/C2=CC=C(O)C(O)=C2)=O)C[C@@H](OC(/C=C/C3=CC=C(O)C(O)=C3)=O)[C@H](O)[C@H](O)C1)O
Molecular Weight (g/mol) 516.45
Formula Weight 516.45
Percent Purity 98.68%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration