Learn More
1,3-Dibromo-2-methoxy-5-nitrobenzene, TRC
Specifications
Specifications
| Chemical Name or Material | 1,3-Dibromo-2-methoxy-5-nitrobenzene |
| CAS | 31106-74-8 |
| Molecular Formula | C7H5Br2NO3 |
| Quantity | 1 g |
| InChI Formula | InChI=1S/C7H5Br2NO3/c1-13-7-5(8)2-4(10(11)12)3-6(7)9/h2-3H,1H3 |
| SMILES | O=N(=O)C1=CC(Br)=C(OC)C(Br)=C1 |
| IUPAC Name | 1,3-dibromo-2-methoxy-5-nitrobenzene |
| Molecular Weight (g/mol) | 310.93 |
| Formula Weight | 308.86 |