Learn More
1,3-Di-o-tolylguanidine, 99%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 176°C to 178°C |
| Quantity | 25 g |
| UN Number | UN2811 |
| Formula Weight | 239.32 |
| Percent Purity | 99% |
| Chemical Name or Material | 1,3-Di-o-tolylguanidine |
Safety and Handling
- 1,3-Di-o-tolylguanidine
- Danger
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 3
- H301-Toxic if swallowed.
- H412-Harmful to aquatic life with long lasting effects.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P330-Rinse mouth.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: 1,3-Di-o-tolylguanidine