Learn More
1,3-Bis(2-methoxyphenoxy)-2-propanol, TRC
Specifications
Specifications
| Chemical Name or Material | 1,3-Bis(2-methoxyphenoxy)propan-2-ol |
| CAS | 16929-60-5 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C17 H20 O5 |
| Quantity | 25 mg |
| Synonym | 1,3-Bis(2-methoxyphenoxy)propan-2-ol, Guaifenesin Imp. D (EP) |
| InChI Formula | InChI=1S/C17H20O5/c1-19-14-7-3-5-9-16(14)21-11-13(18)12-22-17-10-6-4-8-15(17)20-2/h3-10,13,18H,11-12H2,1-2H3 |
| SMILES | COc1ccccc1OCC(O)COc2ccccc2OC |
| IUPAC Name | 1,3-bis(2-methoxyphenoxy)propan-2-ol |
| Molecular Weight (g/mol) | 304.34 |
| Show More |