Learn More
1,3-Bis(2-chloroethyl)urea, TRC
Specifications
Specifications
| Chemical Name or Material | 1,3-bis(2-chloroethyl)urea |
| CAS | 2214-72-4 |
| Molecular Formula | C5 H10 Cl2 N2 O |
| Quantity | 1 g |
| Synonym | Urea, N,N'-bis(2-chloroethyl)-, Urea, 1,3-bis(2-chloroethyl)- (6CI,7CI,8CI), 1,3-Bis(2-chloroethyl)urea, NSC 36198 |
| InChI Formula | InChI=1S/C5H10Cl2N2O/c6-1-3-8-5(10)9-4-2-7/h1-4H2,(H2,8,9,10) |
| SMILES | ClCCNC(=O)NCCCl |
| IUPAC Name | 1,3-bis(2-chloroethyl)urea |
| Molecular Weight (g/mol) | 185.05 |
| Formula Weight | 184.017 |