Learn More
1,3-Benzodioxole-5-glycolic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 1,3-Benzodioxole-5-glycolic Acid |
| CAS | 27738-46-1 |
| Molecular Formula | C9H8O5 |
| Quantity | 500 mg |
| Synonym | alpha-Hydroxy-1,3-benzodioxole-5-acetic Acid, 2-(1,3-Benzodioxol-5-yl)-2-hydroxyacetic Acid, 2-(2H-1,3-Benzodioxol-5-yl)-2-hydroxyacetic Acid, 2-Hydroxy-2-(3,4-methylenedioxyphenyl)acetic Acid, 3,4-(Methylenedioxy)benzeneglycolic Acid, 3,4-(Methylenedioxy)mandelic Acid, 3',4'-Methylenedioxymandelic Acid |
| InChI Formula | InChI=1S/C9H8O5/c10-8(9(11)12)5-1-2-6-7(3-5)14-4-13-6/h1-3,8,10H,4H2,(H,11,12) |
| SMILES | OC(C(=O)O)c1ccc2OCOc2c1 |
| IUPAC Name | 2-(1,3-benzodioxol-5-yl)-2-hydroxyacetic acid |
| Molecular Weight (g/mol) | 196.16 |
| Formula Weight | 196.0372 |