missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Triphenylbenzene, 99+%

Product Code. p-7025713
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
This item is not returnable. View return policy

Product Code. 11479287

Brand: Thermo Scientific Alfa Aesar B21353.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 612-71-5 | C24H18 | 306.41 g/mol

1,3,5-Triphenylbenzene fluorophore as a selective Cu2+ sensor in aqueous media.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3,5-Triphenylbenzene fluorophore as a selective Cu2+ sensor in aqueous media.

Solubility
Solubility in toluene is almost transparent.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 612-71-5
Molecular Formula C24H18
Molecular Weight (g/mol) 306.41
MDL Number MFCD00003060
InChI Key SXWIAEOZZQADEY-UHFFFAOYSA-N
Synonym triphenylbenzene, 5'-phenyl-1,1':3',1-terphenyl, s-triphenylbenzene, m-terphenyl, 5'-phenyl, 1,1':3',1-terphenyl, 5'-phenyl, 5'-phenyl-m-terphenyl, benzene, 1,3,5-triphenyl, 1,1'-biphenyl, 3,5-diphenyl, unii-408mso5wo3, 3,5-diphenyl-1,1'-biphenyl
PubChem CID 11930
IUPAC Name 1,3,5-triphenylbenzene
SMILES C1=CC=C(C=C1)C1=CC(=CC(=C1)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 172°C to 174°C
Boiling Point 460°C
Quantity 50 g
Formula Weight 306.41
Percent Purity ≥99%
Chemical Name or Material 1,3,5-Triphenylbenzene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.