Learn More
1,3,5-Tri(3-pyridyl)-1,5-pentanoate, TRC
Product Code. 30454078
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30454078 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30454078 Supplier Toronto Research Chemicals Supplier No. TRCT82110050MG
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1,3,5-Tri(3-pyridyl)-1,5-pentanoate
CAS 94678-45-2
Molecular Formula C20H17N3O2
Quantity 50 mg
Synonym 1,3,5-Tri-3-pyridinyl-1,5-pentanedione, 1,3,5-Tris(3-pyridyl)pentane-1,5-dione
InChI Formula InChI=1S/C20H17N3O2/c24-20(25)19(17-6-3-11-23-14-17)18(16-5-2-10-22-13-16)8-7-15-4-1-9-21-12-15/h1-14,18-19H,(H,24,25)/b8-7+
SMILES OC(=O)C(C(\C=C\c1cccnc1)c2cccnc2)c3cccnc3
IUPAC Name (E)-2,3,5-tripyridin-3-ylpent-4-enoic acid
Molecular Weight (g/mol) 331.37
Formula Weight 331.1321