Learn More
1-[3,4-Bis(phenylmethoxy)phenyl]ethanone, TRC
Specifications
Specifications
| Chemical Name or Material | 1-[3,4-Bis(phenylmethoxy)phenyl]ethanone |
| CAS | 27628-06-4 |
| Molecular Formula | C22H20O3 |
| Quantity | 25 mg |
| Synonym | 3,4-Bis(benzyloxy)acetophenone, NSC 211430, 3',4'-Bis(benzyloxy)-acetophenone |
| InChI Formula | InChI=1S/C22H20O3/c1-17(23)20-12-13-21(24-15-18-8-4-2-5-9-18)22(14-20)25-16-19-10-6-3-7-11-19/h2-14H,15-16H2,1H3 |
| SMILES | CC(=O)c1ccc(OCc2ccccc2)c(OCc3ccccc3)c1 |
| IUPAC Name | 1-[3,4-bis(phenylmethoxy)phenyl]ethanone |
| Molecular Weight (g/mol) | 332.39 |
| Formula Weight | 332.1412 |