Learn More
1,3:4,6-Di-O-benzylidene-D-mannitol, TRC
Specifications
Specifications
| Chemical Name or Material | 1,3,4,6-Di-O-benzylidene-D-mannitol |
| CAS | 28224-73-9 |
| Molecular Formula | C20 H22 O6 |
| Quantity | 10 g |
| Synonym | 1,34,6-Di-O-Benzylidene-D-Mannitol |
| InChI Formula | InChI=1S/C20H22O6/c21-15-11-23-19(13-7-3-1-4-8-13)25-17(15)18-16(22)12-24-20(26-18)14-9-5-2-6-10-14/h1-10,15-22H,11-12H2/t15-,16-,17-,18-,19?,20?/m1/s1 |
| SMILES | O[C@@H]1COC(O[C@H]1[C@@H]2OC(OC[C@H]2O)c3ccccc3)c4ccccc4 |
| IUPAC Name | (4R,5R)-4-[(4R,5R)-5-hydroxy-2-phenyl-1,3-dioxan-4-yl]-2-phenyl-1,3-dioxan-5-ol |
| Molecular Weight (g/mol) | 358.39 |
| Formula Weight | 358.1416 |