Learn More
1-[2-Hydroxy-4,6-dimethoxy-3-(methoxymethoxy)phenyl]-1-propanone, TRC
Specifications
Specifications
| Chemical Name or Material | 1-[2-Hydroxy-4,6-dimethoxy-3-(methoxymethoxy)phenyl]-1-propanone |
| CAS | 276690-11-0 |
| Molecular Formula | C13H18O6 |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C13H18O6/c1-5-8(14)11-9(17-3)6-10(18-4)13(12(11)15)19-7-16-2/h6,15H,5,7H2,1-4H3 |
| SMILES | CCC(C1=C(OC)C=C(OC)C(OCOC)=C1O)=O |
| IUPAC Name | 1-(2-hydroxy-4,6-dimethoxy-3-(methoxymethoxy)phenyl)propan-1-one |
| Molecular Weight (g/mol) | 270.28 |
| Formula Weight | 270.11 |