Learn More
1,2-Ethane-d4-dithiol, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 1,2-Ethanedithiol-D4 |
| CAS | 100189-81-9 |
| Molecular Formula | C2 D4 H2 S2 |
| Quantity | 0.5 g |
| Synonym | 1,2-Ethane-1,1,2,2-d4-dithiol (9 CI), 1,2-Ethane-d4-dithiol, 1,2-Ethanedithiol-d4, 1,2-Ethanedithiol-D4, Dithioglycol-d4, 1,2-Dimercaptoethane-1,1,2,2-d4 |
| InChI Formula | InChI=1 S/C2H6S2/c3-1-2-4/h3-4 H,1-2H2/i1D2,2D2 |
| SMILES | [2 H]C([2 H])C([2 H])([2 H])S |
| IUPAC Name | 1,1,2,2-tetradeuterioethane-1,2-dithiol |
| Molecular Weight (g/mol) | 98.2237 |
| Formula Weight | 98.0162 g/mol |
| Show More |