Learn More
1,2-Dipalmitoyl-3-O-benzyl-rac-glycerol, TRC
Specifications
Specifications
| Chemical Name or Material | 1,2-Dipalmitoyl-3-O-benzyl-rac-glycerol |
| CAS | 69176-47-2 |
| Molecular Formula | C42 H74 O5 |
| Quantity | 500 mg |
| InChI Formula | InChI=1S/C42H74O5/c1-3-5-7-9-11-13-15-17-19-21-23-25-30-34-41(43)46-38-40(37-45-36-39-32-28-27-29-33-39)47-42(44)35-31-26-24-22-20-18-16-14-12-10-8-6-4-2/h27-29,32-33,40H,3-26,30-31,34-38H2,1-2H3/t40-/m1/s1 |
| SMILES | CCCCCCCCCCCCCCCC(=O)OC[C@@H](COCc1ccccc1)OC(=O)CCCCCCCCCCCCCCC |
| IUPAC Name | [(2R)-2-hexadecanoyloxy-3-phenylmethoxypropyl] hexadecanoate |
| Molecular Weight (g/mol) | 659.034 |
| Formula Weight | 658.554 |