missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dichlorobutane, 97%

Product Code. 11469217
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11469217 5 g 5g
11434240 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11469217 Supplier Thermo Scientific Alfa Aesar Supplier No. B21307.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 616-21-7 | C4H8Cl2 | 127.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 616-21-7
Molecular Formula C4H8Cl2
Molecular Weight (g/mol) 127.01
MDL Number MFCD00013676
InChI Key PQBOTZNYFQWRHU-UHFFFAOYNA-N
Synonym butane, 1,2-dichloro, butane,2-dichloro, 1,2-butylene chloride, acmc-1b1ry, dsstox_cid_7244, dsstox_rid_78366, dsstox_gsid_27244, 1,2-dichlorobutane, pqbotznyfqwrhu-uhfffaoysa
PubChem CID 12017
IUPAC Name 1,2-dichlorobutane
SMILES CCC(Cl)CCl

Specifications

Density 1.112
Boiling Point 124°C
Flash Point 30°C (86°F)
Refractive Index 1.445
Quantity 5 g
UN Number UN1993
Beilstein 1731376
Formula Weight 127.01
Percent Purity ≥97%
Chemical Name or Material 1,2-Dichlorobutane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.