missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dibromo-1,1-dichloroethane, 97%, Thermo Scientific™

Product Code. p-7024325
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11423048 10 g 10g
11433048 50 g 50g
11443048 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11423048 Supplier Thermo Scientific Alfa Aesar Supplier No. A18768.09

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-81-0
Molecular Formula C2H2Br2Cl2
Molecular Weight (g/mol) 256.746
MDL Number MFCD00053228
InChI Key FIYBYNHDEOSJPL-UHFFFAOYSA-N
Synonym ethane, 1,2-dibromo-1,1-dichloro, 1,2-dibromo-2,2-dichloroethane, unii-8d6fn8f1wd, 8d6fn8f1wd, ethane, dibromodichloro, ethane,2-dibromo-1,1-dichloro, 1,2-dibromo-1,1-dichloroethane, 100 microg/ml in methanol
PubChem CID 6401
IUPAC Name 1,2-dibromo-1,1-dichloroethane
SMILES C(C(Cl)(Cl)Br)Br

Specifications

Quantity 10 g
UN Number UN2810
Formula Weight 256.75
Percent Purity 97%
Chemical Name or Material 1,2-Dibromo-1,1-dichloroethane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.