Learn More
1,2-Bismethylenecyclohexane (~75%), TRC
Specifications
Specifications
| Chemical Name or Material | 1,2-Bismethylenecyclohexane |
| CAS | 2819-48-9 |
| Molecular Formula | C8 H12 |
| Quantity | 100 mg |
| Synonym | 1,2-Bis(methylene)cyclohexane, 1,2-Dimethylenecyclohexane, 1,2-Dimethylidenecyclohexane |
| InChI Formula | InChI=1S/C8H12/c1-7-5-3-4-6-8(7)2/h1-6H2 |
| SMILES | C=C1CCCCC1=C |
| IUPAC Name | 1,2-dimethylidenecyclohexane |
| Molecular Weight (g/mol) | 108.18 |
| Formula Weight | 108.0939 |