Learn More
1,2-Bis(3,5-di-tert-butyl-4-hydroxyhydrocinnamoyl)hydrazine, TRC
Specifications
Specifications
| Chemical Name or Material | Irganox 1024 |
| CAS | 32687-78-8 |
| Molecular Formula | C34 H52 N2 O4 |
| Quantity | 10 g |
| Synonym | Hydrazine, 1,2-bis(3,5-di-tert-butyl-4-hydroxyhydrocinnamoyl)- (8CI), 1,2-Bis(3,5-di-tert-butyl-4-hydroxyhydrocinnamoyl)hydrazide, 1,2-Bis(3,5-di-tert-butyl-4-hydroxyhydrocinnamoyl)hydrazine, 1,2-Bis(3,5-tert-butyl-4-hydroxyhydrocinnamoyl)hydrazine, 3,5-Bis(1,1-dimethylethyl)-4-hydroxybenzenepropionic acid 2-[3-[3,5-bis(1,1-dimethylethyl)-4-hydroxyphenyl]-1-oxopropyl]hydrazide, ADK Stab CDA 10, Antioxidant 1024, Antioxidant BPP, CDA 10, IR 1024, Irganox 1024, Irganox GX MD 1024, Irganox MD 1024, Irganox MD 2014, Lowinox MD 24, MD 1024, MD 24, N,N'-Bis[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionyl]hydrazine, N,N'-Bis[3-(3',5'-di-tert-butyl-4'-hydroxyphenyl)propionyl]hydrazine, N,N'-Bis[β-(3,5-di-tert-butyl-4-hydroxyphenyl)propionic acid] hydrazide, N,N'-Bis[β-(3,5-di-tert-butyl-4-hydroxyphenyl)propionyl]hydrazine, Songnox 1024, Songnox 1024FG, Benzenepropanoic acid, 3,5-bis(1,1-dimethylethyl)-4-hydroxy-, 2-[3-[3,5-bis(1,1-dimethylethyl)-4-hydroxyphenyl]-1-oxopropyl]hydrazide |
| InChI Formula | InChI=1S/C34H52N2O4/c1-31(2,3)23-17-21(18-24(29(23)39)32(4,5)6)13-15-27(37)35-36-28(38)16-14-22-19-25(33(7,8)9)30(40)26(20-22)34(10,11)12/h17-20,39-40H,13-16H2,1-12H3,(H,35,37)(H,36,38) |
| SMILES | CC(C)(C)c1cc(CCC(=O)NNC(=O)CCc2cc(c(O)c(c2)C(C)(C)C)C(C)(C)C)cc(c1O)C(C)(C)C |
| IUPAC Name | 3-(3,5-ditert-butyl-4-hydroxyphenyl)-N'-[3-(3,5-ditert-butyl-4-hydroxyphenyl)propanoyl]propanehydrazide |
| Molecular Weight (g/mol) | 552.7877 |
| Formula Weight | 552.3927 |