Learn More
1,2:5,6-Di-O-isopropylidene-alpha-D-ribo-hexofuranose-3-ulose (mixture of keto and ketal forms), TRC
Specifications
Specifications
| Chemical Name or Material | 1,2,5,6-Di-O-isopropylidene-a-D-ribo-hexofuranose-3-ulose |
| CAS | 2847-00-9 |
| Molecular Formula | C12 H18 O6 |
| Quantity | 1 g |
| InChI Formula | InChI=1S/C12H18O6/c1-11(2)14-5-6(16-11)8-7(13)9-10(15-8)18-12(3,4)17-9/h6,8-10H,5H2,1-4H3/t6?,8-,9-,10-/m1/s1 |
| SMILES | CC1(C)OCC(O1)[C@H]2O[C@@H]3OC(C)(C)O[C@@H]3C2=O |
| IUPAC Name | (3aR,5R,6aS)-5-(2,2-dimethyl-1,3-dioxolan-4-yl)-2,2-dimethyl-3a,6a-dihydrofuro[2,3-d][1,3]dioxol-6-one |
| Molecular Weight (g/mol) | 258.27 |
| Formula Weight | 258.1103 |