Learn More
1,2,5,6,9,10-Hexabromocyclododecane, TRC
Product Code. 30396172
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30396172 10 g 10g
30393575 1 g -
30389419 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30396172 Supplier Toronto Research Chemicals Supplier No. TRCH29074010G
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3194-55-6
Molecular Formula C12 H18 Br6
Molecular Weight (g/mol) 641.7
Synonym Cyclododecane, 1,2,5,6,9,10-hexabromo-, 1,2,5,6,9,10-Hexabromocyclododecane, Bromkal 73-6D, FR 1206, FR 1206HT, Pyroguard SR 104, SR 104, YM 88A, Hexabromocyclododecane, HBCD, HBCDD
IUPAC Name 1,2,5,6,9,10-hexabromocyclododecane
SMILES BrC1CCC(Br)C(Br)CCC(Br)C(Br)CCC1Br
Specifications
Quantity 10 g
InChI Formula InChI=1S/C12H18Br6/c13-7-1-2-8(14)10(16)5-6-12(18)11(17)4-3-9(7)15/h7-12H,1-6H2
Formula Weight 635.6509
Chemical Name or Material 1,2,5,6,9,10-Hexabromocyclododecane