Learn More
1-(2,3-Dichlorophenyl)piperazine 1-Oxide, TRC
Specifications
Specifications
| Chemical Name or Material | 1-(2,3-Dichlorophenyl)piperazine 1-Oxide |
| CAS | 902456-05-7 |
| Molecular Formula | C10H12Cl2N2O |
| Quantity | 10 mg |
| InChI Formula | InChI=1S/C10H12Cl2N2O/c11-8-2-1-3-9(10(8)12)14(15)6-4-13-5-7-14/h1-3,13H,4-7H2 |
| SMILES | O=N1(C=2C=CC=C(Cl)C2Cl)CCNCC1 |
| IUPAC Name | 1-(2,3-dichlorophenyl)piperazine 1-oxide |
| Molecular Weight (g/mol) | 247.12 |
| Formula Weight | 246.03 |