Learn More
1,2,3,9-Tetrahydro-9-methyl-4H-carbazol-4-one, TRC
Product Code. 30006553
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006553 100 mg 100mg
30006502 1 g 1g
30004257 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30006553 Supplier Toronto Research Chemicals Supplier No. TRCT293920100MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 27387-31-1
Molecular Formula C13 H13 N O
Molecular Weight (g/mol) 199.25
Synonym 9-Methyl-1,2,3,9-tetrahydro-4H-carbazol-4-one, Ondansetron Hydrochloride Dihydrate Imp. C (EP)
IUPAC Name 9-methyl-2,3-dihydro-1H-carbazol-4-one
SMILES Cn1c2CCCC(=O)c2c3ccccc13
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C13H13NO/c1-14-10-6-3-2-5-9(10)13-11(14)7-4-8-12(13)15/h2-3,5-6H,4,7-8H2,1H3
Formula Weight 199.0997
Percent Purity >95%
Chemical Name or Material 9-Methyl-1,2,3,9-tetrahydro-4H-carbazol-4-one
Grade HPLC