missing translation for 'onlineSavingsMsg'
Learn More

1-[2-(1-Pyrrolidinyl)ethyl]piperazine, 97%, Thermo Scientific™

Product Code. 11490240
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11490240 1 g 1g
11400250 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11490240 Supplier Thermo Scientific Alfa Aesar Supplier No. H50692.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
1-[2-(1-Pyrrolidinyl)ethyl]piperazine is an important raw material and intermediate used in pharmaceuticals, agrochemicals and dyestuff.

Notes
Air Sensitive. Store away from air. Store under dry inert gas. Incompatible with air, carbondioxide and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22763-69-5
Molecular Formula C10H21N3
Molecular Weight (g/mol) 183.299
MDL Number MFCD01075190
InChI Key UXHZIFKPLFIPSX-UHFFFAOYSA-N
Synonym 1-2-pyrrolidin-1-yl ethyl piperazine, 1-2-pyrrolidinoethyl piperazine, 1-2-pyrrolidin-1-ylethyl piperazine, 1-2-pyrrolidin-1-yl-ethyl-piperazine, 1-2-1-pyrrolidinyl ethyl piperazine, piperazine, 1-2-1-pyrrolidinyl ethyl, acmc-1cbwl, 2-pyrrolidinylethyl piperazine, 1-2-pyrrolidinoethyl-piperazine
PubChem CID 2737250
IUPAC Name 1-(2-pyrrolidin-1-ylethyl)piperazine
SMILES C1CCN(C1)CCN2CCNCC2

Specifications

Boiling Point 77°C to 79°C (0.5 mmHg)
Quantity 1 g
UN Number UN2735
Formula Weight 183.3
Percent Purity 97%
Chemical Name or Material 1-[2-(1-Pyrrolidinyl)ethyl]piperazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.