Learn More
1-(1,1-Dimethylpropyl)-4-iodobenzene, TRC
Specifications
Specifications
| Chemical Name or Material | 1-(1,1-Dimethylpropyl)-4-iodobenzene |
| CAS | 860556-27-0 |
| Molecular Formula | C11H15I |
| Quantity | 250 mg |
| Synonym | 1-tert-Amyl-4-iodo-benzene, 1-(1,1-Dimethylpropyl)-4-iodo-benzene |
| InChI Formula | InChI=1S/C11H15I/c1-4-11(2,3)9-5-7-10(12)8-6-9/h5-8H,4H2,1-3H3 |
| SMILES | CCC(C)(C)c1ccc(I)cc1 |
| IUPAC Name | 1-iodo-4-(2-methylbutan-2-yl)benzene |
| Molecular Weight (g/mol) | 274.14 |
| Formula Weight | 274.0218 |