Learn More
Greiner Bio-One 0.7mL Polypropylene Microcentrifuge Tubes
Product Code. 15915361
Change view
Click to view available options
Color:
Blue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color
15915361 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15915361 Supplier Greiner Bio-One Supplier No. 742270
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
For single use only. Greiner Bio-One Cell 0.7mL Polypropylene Microcentrifuge Tube is free of detectable DNase, RNase, human DNA.
  • Material: Polypropylene
  • For single use only
  • With attached cap
  • Free of detectable DNase/RNase, human DNA and pyrogens
  • Non-Pyrogenic
  • High chemical and temperature resistance
  • Suitable for Roche centrifu
Certifications
DIN ISO 9001
TRUSTED_SUSTAINABILITY
Specifications
Color Blue
Material Polypropylene
Volume (Metric) 0.7mL
Sterility Non-sterile