Learn More
Greiner Bio-One 0.7mL Polypropylene Microcentrifuge Tubes
Description
- Material: Polypropylene
- For single use only
- With attached cap
- Free of detectable DNase/RNase, human DNA and pyrogens
- Non-Pyrogenic
- High chemical and temperature resistance
- Suitable for Roche centrifu
Certifications
DIN ISO 9001
Specifications
Specifications
| Color | Blue |
| Material | Polypropylene |
| Volume (Metric) | 0.7mL |
| Sterility | Non-sterile |