missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Butyl-d9-benzene, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | Butylbenzene-D9 (butyl-D9) |
| CAS | 65087-59-4 |
| Molecular Formula | C10 D9 H5 |
| Quantity | 0.5 g |
| Synonym | Benzene, butyl-d9- (9 CI), (n-Butyl-d9)benzene, (Butyl-1,1,2,2,3,3,4,4,4-d9)benzene, Butyl-d9-benzene, Butylbenzene-d9, Butylbenzene-D9 (butyl-D9) |
| InChI Formula | InChI=1 S/C10H14/c1-2-3-7-10-8-5-4-6-9-10/h4-6,8-9 H,2-3,7H2,1H3/i1D3,2D2,3D2,7D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])c1ccccc1 |
| IUPAC Name | 1,1,2,2,3,3,4,4,4-nonadeuteriobutylbenzene |
| Molecular Weight (g/mol) | 143.274 |
| Formula Weight | 143.166 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction