Learn More
Axygen™ 0.5 mL Conical Screw Cap Tubes: Presterilized
Product Code. 12709498 Shop All Axygen Scientific Products
Click to view available options
Closure Color:
Assorted
Clear
Unit Size:
Pack of 4000
Pack of 500
Product Code. 12709498
Brand: Axygen™ SCT050CS
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Features chemical resistant O-ring
Serve a broad range of laboratory needs including collection, processing, centrifugation, storage and packaging/shipping of restriction enzymes and other reagents.
  • Presterilized cap tubes have non-looped caps attached with O-rings
  • Threaded O-ring design ensures uniform closure allowing centrifugation up to 20,000 x g
  • Maxymum Recovery™ screw cap tubes available as special order items for assorted tubes
  • Available in range of colors for easy sample identification
TRUSTED_SUSTAINABILITY
Specifications
Cap Style Screw Cap
Certifications/Compliance RNase, DNase, pyrogen-free
Closure Color Clear
Closure Material Polypropylene
Capacity (Metric) 0.5 mL
Color Clear
Material Polypropylene
Format Non-looped
Includes O-ring
No. per Pack 500/Pk.
Packaging 500 Tubes and caps/Pk.; 8 Packs/Cs.
Sterility Sterile
Graduated No
Closure Type Screw Cap
Bottom Shape Conical
Max. RCF 20,000 x g