missing translation for 'onlineSavingsMsg'
Learn More
Learn More
m-Cresol-d8, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | Metacresol-D8 |
| CAS | 302911-90-6 |
| Molecular Formula | C7 D8 O |
| Quantity | 0.25 g |
| Synonym | Phen-2,3,4,6-d4-ol-d, 5-(methyl-d3)- (9 CI), 3-(Methyl-d3)phenol-hydroxy,2,4,5,6-d5, m-Cresol-d7, 1-(Hydroxy-d)-3-(methyl-d3)benzene-2,4,5,6-d4, 3-Cresol-d8, m-Hydroxytoluene-d8, Metacresol-D8 |
| InChI Formula | InChI=1 S/C7H8O/c1-6-3-2-4-7(8)5-6/h2-5,8 H,1H3/i1D3,2 D,3 D,4 D,5 D/hD |
| SMILES | [2 H]Oc1c([2 H])c([2 H])c([2 H])c(c1[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 1,2,3,5-tetradeuterio-4-deuteriooxy-6-(trideuteriomethyl)benzene |
| Molecular Weight (g/mol) | 116.1871 |
| Formula Weight | 116.1077 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction