missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCP1 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NB300-229CL1
This item is not returnable.
View return policy
Description
SCP1 Polyclonal antibody specifically detects SCP1 in Chicken,Human (Negative),Mammalia,Monkey,Mouse,Parasite,Rat samples. It is validated for ChIP assay,Immunohistochemistry,Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin),Immunohistochemistry (Frozen),Simple Western,ChIP assay (ChIP),Immunocytochemistry/Immunofluorescence
Specifications
| SCP1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Chicken, Human (Negative), Mammalia, Monkey, Mouse | |
| Purified |
| ChIP Assay, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
| CoraFluor 1 | |
| Cancer/testis antigen 8, CT8MGC104417, HOM-TES-14, SCP1SCP-1, synaptonemal complex protein 1 | |
| A synthetic peptide made to the C-terminus of the mouse SCP1 protein sequence. [UniProt# Q62209] | |
| 0.1 mL | |
| Cell Cycle and Replication, Chromatin Research, Growth and Development, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| 6847 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction