missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
GPIHBP1 Polyclonal antibody specifically detects GPIHBP1 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence.
Specifications
Specifications
| Antigen | GPIHBP1 |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Concentration | 0.9 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 2.0 ug/ml, Immunocytochemistry/Immunofluorescence 1:20 - 1:50 |
| Formulation | Tris-Glycine and 0.15M NaCl with 0.05% Sodium Azide |
| Gene Alias | glycosylphosphatidylinositol anchored high density lipoprotein binding protein1, glycosylphosphatidylinositol-anchored high density lipoprotein-binding protein1, GPI anchored high density lipoprotein binding protein 1, GPI-anchored HDL-binding protein 1, GPI-HBP1LOC338328, HBP1, High density lipoprotein-binding protein 1 |
| Gene Symbols | GPIHBP1 |
| Host Species | Rabbit |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?