missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexamethyl-d18-disilane, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | Hexamethyldisilane-D18 |
| CAS | 61123-71-5 |
| Molecular Formula | C6 D18 Si2 |
| Quantity | 0.1 g |
| Synonym | Disilane, hexa(methyl-d3)- (9 CI), 1,1,1,2,2,2-Hexa(methyl-d3)disilane, Hexamethyl-d18-disilane, Hexamethyldisilane-D18, HMD-d18 |
| InChI Formula | InChI=1 S/C6H18Si2/c1-7(2,3)8(4,5)6/h1-6H3/i1D3,2D3,3D3,4D3,5D3,6D3 |
| SMILES | [2 H]C([2 H])([2 H])[Si](C([2 H])([2 H])[2 H])(C([2 H])([2 H])[2 H])[Si](C([2 H])([2 H])[2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | tris(trideuteriomethyl)-[tris(trideuteriomethyl)silyl]silane |
| Molecular Weight (g/mol) | 164.489 |
| Formula Weight | 164.2077 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction