Learn More
Heptadecanoic-17,17,17-d3 Acid, CDN
Product Code. 30320229
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.10g
0.25g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30320229
Brand: CDN CDND52550.1G
Please to purchase this item. Need a web account? Register with us today!
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Heptadecanoic Acid-17,17,17-D3
CAS 202528-95-8
Molecular Formula C17 D3 H31 O2
Quantity 0.1 g
Synonym Heptadecanoic-17,17,17-d3 acid (9 CI), Heptadecanoic Acid-17,17,17-D3, n-Heptadecylic acid-d3, Margaric acid-ω-d3
InChI Formula InChI=1 S/C17H34O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17(18)19/h2-16H2,1H3,(H,18,19)/i1D3
SMILES [2 H]C([2 H])([2 H])CCCCCCCCCCCCCCCC(=O)O
IUPAC Name 17,17,17-trideuterioheptadecanoic acid
Molecular Weight (g/mol) 273.4692
Formula Weight 273.2747 g/mol
Percent Purity 99 atom % D, min 98% Chemical Purity