missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glyceryl Tri(dodecanoate-11,11,12,12,12-d5), CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Glyceryl Tridodecanoate-11,11,11',11',11'',11'',12,12,12,12',12',12',12'',12'',12''-D15 |
| CAS | 1219805-25-0 |
| Molecular Formula | C39 D15 H59 O6 |
| Quantity | 0.1 g |
| Synonym | Glyceryl Tri(dodecanoate-11,11,12,12,12-d5), Dodecanoic-11,11,12,12,12-d5 acid 1,2,3-propanetriyl ester, 1,1',1''-(1,2,3-Propanetriyl) tri dodecanoate-11,11,12,12,12-d5, 1,2,3-Propanetriol tris(dodecanoate-11,11,12,12,12-d5), Glyceryl Tri(dodecanoate-11,11,12,12,12-d5), Glyceryl Tridodecanoate-11,11,11',11',11'',11'',12,12,12,12',12',12',12'',12'',12''-D15, Glyceryl Tridodecanoate-11,11,11',11',11'',11'',12,12,12,12',12',12',12'',12'',12''-D15 |
| InChI Formula | InChI=1 S/C39H74O6/c1-4-7-10-13-16-19-22-25-28-31-37(40)43-34-36(45-39(42)33-30-27-24-21-18-15-12-9-6-3)35-44-38(41)32-29-26-23-20-17-14-11-8-5-2/h36H,4-35H2,1-3H3/i1D3,2D3,3D3,4D2,5D2,6D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])CCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCC([2 H])([2 H])C([2 H])([2 H])[2 H])OC(=O)CCCCCCCCCC([2 H])([2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 2,3-bis(11,11,12,12,12-pentadeuteriododecanoyloxy)propyl 11,11,12,12,12-pentadeuteriododecanoate |
| Molecular Weight (g/mol) | 654.0937 |
| Formula Weight | 653.6427 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction