missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Phenoxy-d5-benzoic-d4 Acid, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.1g
Specifications
Specifications
| Chemical Name or Material | 3-Phenoxybenzoic Acid-D9 |
| Molecular Formula | C13 D9 H O3 |
| Quantity | 0.1 g |
| Synonym | 3-Phenoxy-d5-benzoic-d4 Acid, 5-(Phenoxy-d5)benzoic-2,3,4,6-d4 Acid, 3-Phenoxybenzoic Acid-D9 |
| InChI Formula | InChI=1 S/C13H10O3/c14-13(15)10-5-4-8-12(9-10)16-11-6-2-1-3-7-11/h1-9 H,(H,14,15)/i1D,2 D,3 D,4 D,5 D,6 D,7 D,8 D,9 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(Oc2c([2 H])c([2 H])c([2 H])c(C(=O)O)c2[2 H])c([2 H])c1[2 H] |
| IUPAC Name | 2,3,4,6-tetradeuterio-5-(2,3,4,5,6-pentadeuteriophenoxy)benzoic acid |
| Molecular Weight (g/mol) | 223.2722 |
| Formula Weight | 223.1195 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction