missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl-d6 Chlorothiophosphate, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Dimethyl Chlorothiophosphate-D6 |
| CAS | 19262-97-6 |
| Molecular Formula | C2 D6 Cl O2 P S |
| Quantity | 0.05 g |
| InChI Formula | InChI=1 S/C2H6ClO2PS/c1-4-6(3,7)5-2/h1-2H3/i1D3,2D3 |
| SMILES | [2 H]C([2 H])([2 H])OP(=S)(Cl)OC([2 H])([2 H])[2 H] |
| IUPAC Name | chloro-sulfanylidene-bis(trideuteriomethoxy)-λ5-phosphane |
| Molecular Weight (g/mol) | 166.597 |
| Formula Weight | 165.989 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction