missing translation for 'onlineSavingsMsg'
Learn More

Dichlorvos-d6 (dimethyl-d6), CDN

Product Code. 30320636
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30320636

Brand: CDN CDND38470.05G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dichlorvos-d6
CAS 203645-53-8
Molecular Formula C4 2H6 H Cl2 O4 P
Quantity 0.05 g
Synonym Phosphoric acid, 2,2-dichloroethenyl di(methyl-d3) ester, 2,2-Dichloroethenyl di(methyl-d3) phosphate, Dichlorvos D6 (dimethyl D6), Dichlorvos-d6
InChI Formula InChI=1 S/C4H7Cl2O4P/c1-8-11(7,9-2)10-3-4(5)6/h3H,1-2H3/i1D3,2D3
SMILES [2 H]C([2 H])([2 H])OP(=O)(OC=C(Cl)Cl)OC([2 H])([2 H])[2 H]
IUPAC Name 2,2-dichloroethenyl bis(trideuteriomethyl) phosphate
Molecular Weight (g/mol) 227.01
Formula Weight 225.9836 g/mol
Percent Purity 99 atom % D, min 95% Chemical Purity
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.