missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Piroxicam-d4 (pyridyl-d4), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Piroxicam-D4 (pyridinyl-D4) |
| Molecular Formula | C15 D4 H9 N3 O4 S |
| Quantity | 0.01 g |
| Synonym | Piroxicam D4 (pyridinyl D4), Piroxicam-D4 (pyridinyl-D4), 4-Hydroxy-2-methyl-N-(2-pyridinyl-3,4,5,6-d4)-2 H-1,2-benzothiazine-3-carboxamide 1,1-dioxide, 4-Hydroxy-2-methyl-N-(2-pyridyl-d4)-2 H-1,2-benzothiazine-3-carboxamide 1,1-dioxide, Piroxicam-d4, Piroxicam-d4 (pyridyl-d4) |
| InChI Formula | InChI=1 S/C15H13N3O4S/c1-18-13(15(20)17-12-8-4-5-9-16-12)14(19)10-6-2-3-7-11(10)23(18,21)22/h2-9,19 H,1H3,(H,16,17,20)/i4D,5 D,8 D,9 D |
| SMILES | [2 H]c1nc(NC(=O)C2=C(O)c3ccccc3S(=O)(=O)N2C)c([2 H])c([2 H])c1[2 H] |
| IUPAC Name | 4-hydroxy-2-methyl-1,1-dioxo-N-(3,4,5,6-tetradeuteriopyridin-2-yl)-1λ^{6},2-benzothiazine-3-carboxamide |
| Molecular Weight (g/mol) | 335.371 |
| Formula Weight | 335.088 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction