missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ciprofloxacin-d8 HCl H2O (piperazine-d8), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Ciprofloxacin-D8 (piperazine-D8) Hydrochloride Monohydrate |
| Molecular Formula | C17 D8 H10 F N3 O3 . Cl H . H2 O |
| Quantity | 0.01 g |
| Synonym | 1-Cyclopropyl-6-fluoro-7-(2,2,3,3,5,5,6,6-octadeuteriopiperazin-1-yl)-4-oxoquinoline-3-carboxylic acid hydrochloride monohydrate, 1-Cyclopropyl-6-fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl-2,2,3,3,5,5,6,6-d8)-3-quinolinecarboxylic acid hydrochloride hydrate (1:1:1), Ciprofloxacin-d8 hydrochloride monohydrate, Ciprofloxacin-D8 (piperazine-D8) hydrochloride monohydrate, Ciprofloxacin-d8 HCl H2O (piperazine-d8) |
| InChI Formula | InChI=1 S/C17H18FN3O3.ClH.H2O/c18-13-7-11-14(8-15(13)20-5-3-19-4-6-20)21(10-1-2-10)9-12(16(11)22)17(23)24;;/h7-10,19 H,1-6H2,(H,23,24);1 H;1H2/i3D2,4D2,5D2,6D2;; |
| SMILES | O.Cl.[2 H]C1([2 H])NC([2 H])([2 H])C([2 H])([2 H])N(c2cc3N(C=C(C(=O)O)C(=O)c3cc2F)C4CC4)C1([2 H])[2 H] |
| IUPAC Name | 1-cyclopropyl-6-fluoro-7-(2,2,3,3,5,5,6,6-octadeuteriopiperazin-1-yl)-4-oxoquinoline-3-carboxylic acid;hydrate;hydrochloride |
| Molecular Weight (g/mol) | 393.867 |
| Formula Weight | 393.1707 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction