missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbamazepine-d10 (rings-d10), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Carbamazepine-D10 |
| CAS | 132183-78-9 |
| Molecular Formula | C15 2H10 H2 N2 O |
| Quantity | 0.005 g |
| Synonym | Carbamazepine-D10, 5 H-Dibenz[b,f]azepine-1,2,3,4,6,7,8,9,10,11-d10-5-carboxamide |
| InChI Formula | InChI=1 S/C15H12N2O/c16-15(18)17-13-7-3-1-5-11(13)9-10-12-6-2-4-8-14(12)17/h1-10 H,(H2,16,18)/i1D,2 D,3 D,4 D,5 D,6 D,7 D,8 D,9 D,10 D |
| SMILES | [2 H]C1=C([2 H])c2c([2 H])c([2 H])c([2 H])c([2 H])c2N(C(=O)N)c3c([2 H])c([2 H])c([2 H])c([2 H])c13 |
| IUPAC Name | 1,2,3,4,5,6,7,8,9,10-decadeuteriobenzo[b][1 ]benzazepine-11-carboxamide |
| Molecular Weight (g/mol) | 246.33 |
| Formula Weight | 246.1577 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction