missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-Carazolol-d7 (iso-propyl-d7), CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Carazolol D7 |
| CAS | 1173021-02-7 |
| Molecular Formula | C18 2H7 H15 N2 O2 |
| Quantity | 0.01 g |
| Synonym | Carazolol D7 |
| InChI Formula | InChI=1 S/C18H22N2O2/c1-12(2)19-10-13(21)11-22-17-9-5-8-16-18(17)14-6-3-4-7-15(14)20-16/h3-9,12-13,19-21 H,10-11H2,1-2H3/i1D3,2D3,12 D |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])(NCC(O)COc1cccc2[nH]c3ccccc3c12)C([2 H])([2 H])[2 H] |
| IUPAC Name | 1-(9 H-carbazol-4-yloxy)-3-(1,1,1,2,3,3,3-heptadeuteriopropan-2-ylamino)propan-2-ol |
| Molecular Weight (g/mol) | 305.42 |
| Formula Weight | 305.2121 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction