Learn More
(±)-Amlodipine-d4 Maleate, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Amlodipine-d4 Maleic Acid Salt (2-Chlorophenyl-d4) |
| CAS | 2714486-25-4 |
| Molecular Formula | C20 D4 H21 Cl N2 O5 . C4 H4 O4 |
| Quantity | 0.005 g |
| Synonym | (±)-Amlodipine-d4 Maleate, Amlodipine-d4 maleate, Amlodipine-D4 (2-chlorophenyl-D4) Maleate, 2-[(2-Aminoethoxy)methyl]-4-(2-chlorophenyl-3,4,5,6-d4)-3-(ethoxycarbonyl)-5-(methoxycarbonyl)-6-methyl-1,4-dihydropyridine (2 Z)-2-butenedioate (1:1), 2-[(2-Aminoethoxy)methyl]-4-(2-chlorophenyl-d4)-1,4-dihydro-6-methyl-3,5-pyridinedicarboxylic acid 3-ethyl 5-methyl ester (2 Z)-2-butenedioate |
| InChI Formula | InChI=1 S/C20H25ClN2O5.C4H4O4/c1-4-28-20(25)18-15(11-27-10-9-22)23-12(2)16(19(24)26-3)17(18)13-7-5-6-8-14(13)21;5-3(6)1-2-4(7)8/h5-8,17,23 H,4,9-11,22H2,1-3H3;1-2 H,(H,5,6)(H,7,8)/b;2-1-/i5D,6 D,7 D,8 D; |
| SMILES | [2 H]c1c([2 H])c([2 H])c(C2C(=C(C)NC(=C2C(=O)OCC)COCCN)C(=O)OC)c(Cl)c1[2 H].OC(=O)\C=C/C(=O)O |
| IUPAC Name | (Z)-but-2-enedioic acid;3-O-ethyl 5-O-methyl 2-(2-aminoethoxymethyl)-4-(2-chloro-3,4,5,6-tetradeuteriophenyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate |
| Molecular Weight (g/mol) | 528.9727 |
| Formula Weight | 528.1813 g/mol |
| Show More |