SureTrace Program Featured Product
Contec - a SureTRACE  Program Supplier Partner.
Contec™ Filtered Alcohols
70% IPA or Denatured Ethanol Blended with Purified Water.
Ensures alcohol, container, packaging are cleanroom free from contamination and particulates.
  • GMP manufactured under Grade A air flow in a Grade C cleanroom
  • Ensures the alcohol, container and packaging are free from contamination and particulates
  • No denaturants other than IPA
  • Completely residue free
  • Trigger sprayer and bag in bottle protected system
  • Prevents bottle contents from becoming contaminated during use
  • No need to discard unused product during shelf life
  • Bottles can be completely emptied eliminating wastage
  • Trigger sprayer can be set to jet or spray
  • Large droplet size reduces the risk of inhalation and provides good surface coverage
  • Double bagged packed in linear tear packaging
  • Each bag is easy to open even when wearing gloves
  • Facilitates transfer disinfection into cleanroom