Rotary Evaporators
Rotary Evaporators
Rotary evaporators are essential for efficient and gentle removal of solvents in chemistry, pharma and life science laboratories. From routine distillations to complex synthesis workflows, they support safe concentration, drying and solvent recycling with precise control of rotation, temperature and vacuum.
Cole-Parmer™ Stuart™ Rotary evaporator, vertical coil condenser
Compact Digital Rotary evaporator, with Vertical glassware set, ideal for solvent extraction where there is limited bench space.
Heidolph™ Condenser G1
Spare condenser for rotary evaporator. Heidolph™ Condenser G1 is designed for the LABORATA series of evaporators.
IKA™ RV 8 Rotary Evaporator with Dry Ice Condenser
The RV 8 Rotary Evaporator with Dry Ice Condenser offers a new distilling system with a manual lift and new clamp mechanism. 
Save Now - Exclusive Deals
FAQ
A rotary evaporator gently removes volatile solvents by rotating a flask in a heated bath under reduced pressure. The solvent vapour is condensed and collected for reuse or disposal. Systems from Buchi, Heidolph, IKA and Stuart combine controlled heating, rotation and vacuum for efficient and reproducible solvent removal.
Start by defining solvent type, boiling point, sample volume and required throughput. Then select a rotary evaporator with suitable bath temperature, condenser style and flask capacity. Buchi, Heidolph, IKA and Stuart offer scalable platforms from teaching labs to process development, helping you match performance and budget.
Key accessories include compatible condensers, vacuum pumps, recirculating chillers, receiving flasks and protective coatings. IKA and Heidolph offer matched condensers and starter solutions, while Stuart and Buchi provide dedicated glass sets. Choosing matched accessories improves efficiency, safety and serviceability across your fleet.
Fisher Scientific combines leading brands with EU-wide product availability and consistent contract pricing. Rotary evaporators and accessories from Buchi, Heidolph, IKA and Stuart can be standardised across sites, simplifying qualification, training and maintenance while leveraging centralised procurement and inventory control.
Top Brands