accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 99747-74-7

FFFSOOO

CAS RN 99747-74-7

IUPAC Name: naphthalen-1-yl trifluoromethanesulfonate
Synonyms: naphthalen-1-yl trifluoromethanesulfonate
Molecular Weight (g/mol): 276.23
Molecular Formula: C11H7F3O3S
InChi Key: WQWUQDVFRYMMCY-UHFFFAOYSA-N
SMILES: FC(F)(F)S(=O)(=O)OC1=C2C=CC=CC2=CC=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results