accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 99420-15-2

ONSN

CAS RN 99420-15-2

IUPAC Name: 2-(pyridin-2-yl)-4H-1,3-benzothiazin-4-one
Synonyms: 2-(pyridin-2-yl)-1,3-benzothiazin-4-one
Molecular Weight (g/mol): 240.28
Molecular Formula: C13H8N2OS
InChi Key: GBAKVEWPYUIGHN-UHFFFAOYSA-N
SMILES: O=C1N=C(SC2=CC=CC=C12)C1=CC=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results