accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 992-92-7

TiCH3OCH3OCH3OCH3O

CAS RN 992-92-7

IUPAC Name: titanium(4+) tetramethanolate
Synonyms: titanium(4+) tetrakis(methoxide)
Molecular Weight (g/mol): 172
Molecular Formula: C4H12O4Ti
InChi Key: ITNVWQNWHXEMNS-UHFFFAOYSA-N
SMILES: [Ti+4].C[O-].C[O-].C[O-].C[O-]
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)